(1R,2S)-2-Aminocyclopentanol hydrochloride 95%

Supplier: Ambeed
Warning

A219695-100MG A219695-1G A219695-250MG A219695-25G A219695-5G
76691-756EA 15.92 USD
76691-756 76691-758 76691-760 76691-762 76691-764
(1R,2S)-2-Aminocyclopentanol hydrochloride 95%
(1R,2S)-2-Aminocyclopentanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO·ClH
MW: 137.608 g/mol
MDL Number: MFCD07370091
CAS Number: 137254-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR