Ammonium trifluoroacetate 98%

Supplier: Ambeed
Warning

Synonyms: Ammoniumtrifluoroacetate, Trifluoroacetic acid ammonium salt

A219601-100G A219601-25G A219601-5G
76691-750EA 58 USD
76691-750 76691-752 76691-754
Ammonium trifluoroacetate 98%
Ammonium trifluoroacetate

Precaución: for Research Use Only

Formula: C₂H₄F₃NO₂
MW: 131.05
MDL Number: MFCD00012615
CAS Number: 3336-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR