N,N'-Bis(salicylidene)-1,2-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: N,N'-Bis(2-hydroxybenzylidene)-1,2-phenylenediamine, N,N'-Disalicylal-1,2-phenylenediamine, 2,2'-((1,2-Phenylenebis(azanylylidene))bis(methanylylidene))diphenol

A219353-100G A219353-1G A219353-25G A219353-5G
76691-740EA 435 USD
76691-740 76691-742 76691-744 76691-746
N,N'-Bis(salicylidene)-1,2-phenylenediamine 97%
N,N'-Bis(salicylidene)-1,2-phenylenediamine

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂O₂
MDL Number: MFCD00009995
CAS Number: 3946-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR