Calcium acetylacetonate hydrate 98% + (16-18%Ca)

Supplier: Ambeed
Warning

Synonyms: Bis(2,4-pentanedionato)calcium(II), Acetylacetone Calcium(II) Salt, Calcium(II) Acetylacetonate, Calciumacetylacetonate

A219044-1000G A219044-100G A219044-500G
76691-728EA 23 USD
76691-728 76691-730 76691-732
Calcium acetylacetonate hydrate 98% + (16-18%Ca)
Calcium acetylacetonate hydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₄CaO₄·xH₂O
MW: 238.30 g/mol (anhy)
MDL Number: MFCD00150163
CAS Number: 19372-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR