Allyl-ɑ-D-galactopyranoside 98%

Supplier: Ambeed
Warning

Synonyms: Ally a-D-galactopyranoside, Allyl α-D-Galactopyranoside, 2-Propen-1-yl α-D-Galactopyranoside, α-D-Galactose Monoallyl Ether

A218417-1G A218417-250MG A218417-5G A218417-10G
76691-712EA 48 USD
76691-712 76691-714 76691-716 77877-683
Allyl-ɑ-D-galactopyranoside 98%
Allyl-alpha-D-galactopyranoside

Precaución: for Research Use Only

Formula: C₉H₁₆O₆
MDL Number: MFCD01320363
CAS Number: 48149-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR