Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate

A218166-1G A218166-5G A218166-250MG A218166-100MG
76691-708EA 160 USD
76691-708 76691-710 77593-420 77626-106
Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 98%
Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate

Precaución: for Research Use Only

Formula: C₂₀H₂₀Cl₂N₄O₂Ru
MW: 520.37 g/mol
MDL Number: MFCD00015504
CAS Number: 609844-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR