Azilsartan 98%

Supplier: Ambeed
Warning

A217890-100MG A217890-1G A217890-250MG A217890-25G A217890-50MG A217890-5G A217890-10MG
76691-684EA 8 USD
76691-684 76691-686 76691-688 76691-690 76691-692 76691-694 77626-104
Azilsartan 98%
Azilsartan

Precaución: for Research Use Only

Formula: C₂₅H₂₀N₄O₅
MDL Number: MFCD20278186
CAS Number: 147403-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR