2-Methyl-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-(o-tolyl)propanoic acid, H-D-Phe(2-Me)-OH

A217346-1G A217346-250MG A217346-5G
76691-672EA 36.08 USD
76691-672 76691-674 76691-676
2-Methyl-D-phenylalanine 97%
2-Methyl-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.219 g/mol
MDL Number: MFCD01860870
CAS Number: 80126-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR