D-Alanine benzyl ester-p-toluenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: D-Alanine benzyl ester p-toluenesulphonate salt, H-D-Ala-OBzl.TosOH, D-Alanine Benzyl Ester p-Toluenesulphonate H-D-Ala-OBzl·TosOH D-Ala-OBzl·Tos, H-D-Ala-OBzl·TosOH, H-D-Ala-OBzl.Tos-OH

A217019-1G A217019-5G A217019-10G A217019-25G
76691-660EA 11.67 USD
76691-660 76691-662 77626-096 77626-098
D-Alanine benzyl ester-p-toluenesulfonate 95%
D-Alanine benzyl ester-p-toluenesulphonate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 351.42 g/mol
MDL Number: MFCD00039092
CAS Number: 41036-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR