4,5-Dimethoxy-2-iodobenzoic acid 95%

Supplier: Ambeed
Warning

A216534-100G A216534-10G A216534-1G A216534-250MG A216534-25G A216534-5G
76691-648EA 649 USD
76691-648 76691-650 76691-652 76691-654 76691-656 76691-658
4,5-Dimethoxy-2-iodobenzoic acid 95%
4,5-Dimethoxy-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉IO₄
MDL Number: MFCD00079751
CAS Number: 61203-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR