4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (4-(1-Phenyl-1H-benzo[d]imidazol-2-yl)phenyl)boronic acid, 4-(1-Phenyl-1H-benzimidazol-2-yl)benzeneboronic Acid

A215947-100G A215947-1G A215947-25G A215947-5G
76691-620EA 419 USD
76691-620 76691-622 76691-624 76691-626 76691-628
4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid 96%
4-(1-Phenyl-1H-benzimidazol-2-yl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₅BN₂O₂
MDL Number: MFCD11045057
CAS Number: 952514-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR