Homovanillic acid 95%

Supplier: Ambeed
Warning

A215667-100G A215667-1G A215667-25G A215667-5G
76691-606EA 308 USD
76691-606 76691-608 76691-610 76691-612
Homovanillic acid 95%
Homovanillic acid

Precaución: for Research Use Only

Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MDL Number: MFCD00004350
CAS Number: 306-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR