cis-4-Cyclohexene-1,2-dicarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-rel-Cyclohex-4-ene-1,2-dicarboxylic acid

A214094-100G A214094-500G A214094-25G
76691-584EA 17 USD
76691-584 77626-078 77879-319
cis-4-Cyclohexene-1,2-dicarboxylic acid 98%
cis-4-Cyclohexene-1,2-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MW: 170.17 g/mol
MDL Number: MFCD00070482
CAS Number: 2305-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR