4-Nitro-2,6-xylenol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-nitro-3-xylene, 2,6-Dimethyl-4-nitrophenol

A213449-1G A213449-25G A213449-500G A213449-5G A213449-10G
76691-562EA 6.37 USD
76691-562 76691-564 76691-566 76691-568 77626-076
4-Nitro-2,6-xylenol 98%
4-Nitro-2,6-xylenol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007339
CAS Number: 2423-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR