(+)-Biotin-4-nitrophenyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrophenyl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin-ONP

A212345-100MG A212345-1G A212345-250MG
76691-546EA 21.22 USD
76691-546 76691-548 76691-550
(+)-Biotin-4-nitrophenyl ester 98%
(+)-Biotin-4-nitrophenyl ester

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₅S
MW: 365.40 g/mol
MDL Number: MFCD15141972
CAS Number: 33755-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR