Benzyl 4-bromobutanoate 95%

Supplier: Ambeed
Warning

A212163-100G A212163-100MG A212163-1G A212163-250MG A212163-25G A212163-5G
76691-530EA 379 USD
76691-530 76691-532 76691-534 76691-536 76691-538 76691-540
Benzyl 4-bromobutanoate 95%
Benzyl 4-bromobutanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MDL Number: MFCD00143681
CAS Number: 126430-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR