4-Propoxyphenol 97%

Supplier: Ambeed
Warning

A211520-10G A211520-250MG A211520-5G A211520-25G A211520-1G
76691-508EA 37 USD
76691-508 76691-510 76691-512 77626-070 77877-663
4-Propoxyphenol 97%
4-Propoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00002335
CAS Number: 18979-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR