(1R)-(-)-Camphor-10-sulphonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: (1R)-(-)-2-Oxobornane-10-sulphonyl chloride, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonyl chloride, (1R)-(-)-Camphor-10-sulphonic acid chloride, L(-)-Camphor-10-sulphonyl chloride

A211302-1G A211302-25G A211302-5G
76691-496EA 10 USD
76691-496 76691-498 76691-500
(1R)-(-)-Camphor-10-sulphonyl chloride 97%
(1R)-(-)-Camphor-10-sulphonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClO₃S
MDL Number: MFCD00064155
CAS Number: 39262-22-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR