Cyclobenzaprine hydrochloride 99%

Supplier: Ambeed
Danger

Synonyms: 3-(5H-Dibenzo[a,d][7]annulen-5-ylidene)-N,N-dimethylpropan-1-amine hydrochloride, Cyclobenzaprine HCI

A210756-100MG A210756-1G A210756-1MG A210756-250MG A210756-5G A210756-25G
76691-476EA 13 USD
76691-476 76691-478 76691-480 76691-482 77626-068 77879-308
Cyclobenzaprine hydrochloride 99%
Cyclobenzaprine hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₁N·HCl
MDL Number: MFCD00079039
CAS Number: 6202-23-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR