(3-Fluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Fluorophenyl)acetic acid

A210628-100G A210628-10G A210628-500G A210628-25G
76691-454EA 45 USD
76691-454 76691-456 76691-458 76691-460 77087-102
(3-Fluorophenyl)acetic acid 97%
(3-Fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄F)CH₂COOH
MDL Number: MFCD00004331
CAS Number: 331-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR