cis-Platin 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride
A210558-100MG
A210558-1G
A210558-1MG
A210558-250MG
76691-448EA
18.04
USD
76691-448
76691-450
76691-452
77568-032
cis-Platin 99%
cis-Platin
Precaución: For research use only.
Formula:
H₆Cl₂N₂Pt MW: 300.05 g/mol |
MDL Number:
MFCD00011623 CAS Number: 15663-27-1 UN: 3288 ADR: 6.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...