3-(Trifluoromethyl)-D-phenylalanine 98%

Supplier: Ambeed
Warning

A209893-100MG A209893-1G A209893-250MG A209893-5G
76691-426EA 7 USD
76691-426 76691-428 76691-430 76691-432
3-(Trifluoromethyl)-D-phenylalanine 98%
3-(Trifluoromethyl)-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
MDL Number: MFCD01860875
CAS Number: 14464-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR