Methyl-2-amino-4,5-dimethoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4,5-dimethoxyanthranilate, 2-Amino-4,5-dimethoxy benzoate, Methyl 2-amino-4,5-dimethoxybenzoate, 2-Amino-4,5-dimethoxybenzoic acid methyl ester

A209216-100G A209216-10G A209216-25G A209216-500G A209216-5G
76691-406EA 54.11 USD
76691-406 76691-408 76691-410 76691-412 76691-414
Methyl-2-amino-4,5-dimethoxybenzoate 98%
Methyl-2-amino-4,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
MDL Number: MFCD00014904
CAS Number: 26759-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR