Methyl 3-amino-5-chloro-2-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A209011-1G A209011-250MG
76691-402EA 51.99 USD
76691-402 76691-404
Methyl 3-amino-5-chloro-2-hydroxybenzoate 98%
Methyl 3-amino-5-chloro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
MDL Number: MFCD03768414
CAS Number: 5043-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR