8-Bromo-2-chloroquinazoline 97%

Supplier: Ambeed
Warning

A208719-100MG A208719-10G A208719-1G A208719-250MG A208719-25G A208719-5G
76691-382EA 23.34 USD
76691-382 76691-384 76691-386 76691-388 76691-390 76691-392
8-Bromo-2-chloroquinazoline 97%
8-Bromo-2-chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD11043845
CAS Number: 956100-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR