Anethole 98%

Supplier: Ambeed
Warning

Synonyms: cis-Anethol, 1-Methoxy-4-(prop-1-en-1-yl)benzene

A208633-100G A208633-10G A208633-25G A208633-500G
76691-374EA 21.22 USD
76691-374 76691-376 76691-378 76691-380
Anethole 98%
Anethole

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00009284
CAS Number: 104-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR