9,9'-Bianthracene 97%

Supplier: Ambeed
Warning

Synonyms: 9,9'-Bianthracenyl, 9,9'-Bianthryl

A208432-1G A208432-250MG A208432-5G
76691-368EA 91.24 USD
76691-368 76691-370 76691-372
9,9'-Bianthracene 97%
9,9'-Bianthracene

Precaución: for Research Use Only

Formula: C₂₈H₁₈
MW: 354.45 g/mol
MDL Number: MFCD00130216
CAS Number: 1055-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR