7-Methoxyflavone 98%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-2-phenyl-4H-1-benzopyran-4-one, 7-Methoxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Methoxy-2-phenylchromen-4-one, 7-Methoxy-2-phenyl-4H-chromen-4-one, 7-Methoxy-2-phenyl-4-benzopyrone

A208311-10G A208311-25G A208311-5G
76691-362EA 56.23 USD
76691-362 76691-364 76691-366
7-Methoxyflavone 98%
7-Methoxyflavone

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₃
MW: 252.27 g/mol
MDL Number: MFCD00017462
CAS Number: 22395-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR