D-3-Bromocamphor-10-sulfonic acid monohydrate 98%

Supplier: Ambeed
Danger

A207012-10G A207012-1G A207012-250MG A207012-25G A207012-5G
76691-334EA 147 USD
76691-334 76691-336 76691-338 76691-340 76691-342
D-3-Bromocamphor-10-sulfonic acid monohydrate 98%
D-3-Bromocamphor-10-sulfonic acid monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₅BrO₄S·H₂O
MDL Number: MFCD00209633
CAS Number: 209736-59-4
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR