Sodium anthraquinone-1-sulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulfonate
, Sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulphonate
, Anthraquinone-1-sulfonic acid Sodium Salt
, Anthraquinone-1-sulphonic acid Sodium Salt
A206474-1G
76691-326EA
7.21
USD
76691-326
Sodium anthraquinone-1-sulfonate 98%
Sodium anthraquinone-1-sulfonate
Precaución: for Research Use Only
Formula:
C₁₄H₈O₅S MW: 310.25 g/mol |
MDL Number:
MFCD00037145 CAS Number: 128-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...