1-(4-Chlorophenylsulfonyl)-3-propylurea 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-N-(propylcarbamoyl)benzenesulphonamide, Chlorpropamide

A205674-100G A205674-100MG A205674-10G A205674-25G A205674-5G
76691-316EA 70.02 USD
76691-316 76691-318 76691-320 76691-322 76691-324
1-(4-Chlorophenylsulfonyl)-3-propylurea 98%
1-(4-Chlorophenylsulfonyl)-3-propylurea

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O₃S
MW: 276.74 g/mol
MDL Number: MFCD00079004
CAS Number: 94-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR