Dibenzo[b,d]thiophene 5-oxide 95%

Supplier: Ambeed
Warning

A205475-1G A205475-250MG A205475-5G A205475-25G
76691-310EA 24 USD
76691-310 76691-312 76691-314 77600-674
Dibenzo[b,d]thiophene 5-oxide 95%
Dibenzo[b,d]thiophene 5-oxide

Precaución: for Research Use Only

Formula: C₁₂H₈OS
MDL Number: MFCD00046902
CAS Number: 1013-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR