(3aS,6R)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 98%

Supplier: Ambeed
Warning

A204918-25G A204918-5G
76691-306EA 172.93 USD
76691-306 76691-308
(3aS,6R)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 98%
(3aS,6R)-8,8-Dimethyl-4,5,6,7-tetrahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.3 g/mol
MDL Number: MFCD09992876
CAS Number: 60886-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR