4'-Fluoro-2'-hydroxyacetophenone 98%
Supplier: Ambeed
|
Warning
|
A203976-100G
A203976-10G
A203976-1G
A203976-25G
A203976-5G
A203976-500G
76691-292EA
167
USD
76691-292
76691-294
76691-296
76691-298
76691-300
77877-633
4'-Fluoro-2'-hydroxyacetophenone 98%
4'-Fluoro-2'-hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇FO₂ |
MDL Number:
MFCD00143203 CAS Number: 1481-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...