(3-Bromophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A202848-100G A202848-10G A202848-25G A202848-500G A202848-5G
76691-254EA 59 USD
76691-254 76691-256 76691-258 76691-260 76691-262
(3-Bromophenyl)acetic acid 98%
(3-Bromophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄Br)CH₂COOH
MDL Number: MFCD00004330
CAS Number: 1878-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR