Bis(4-bromophenyl)amine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dibromodiphenylamine

A202168-100G A202168-10G A202168-1G A202168-25G A202168-5G
76691-236EA 55 USD
76691-236 76691-238 76691-240 76691-242 76691-244
Bis(4-bromophenyl)amine 97%
Bis(4-bromophenyl)amine

Precaución: for Research Use Only

Formula: C₁₂H₉Br₂N
MDL Number: MFCD00225488
CAS Number: 16292-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR