5-Isoquinolinol 97%

Supplier: Ambeed
Warning

A202040-100G A202040-10G A202040-1G A202040-250MG A202040-25G A202040-5G
76691-216EA 262 USD
76691-216 76691-218 76691-220 76691-222 76691-224 76691-226
5-Isoquinolinol 97%
5-Isoquinolinol

Precaución: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00006906
CAS Number: 2439-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR