Boc-Lys(Z)-OSu 96%

Supplier: Ambeed
Warning

Synonyms: Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine N-succinimidyl ester

A202006-1G A202006-10G A202006-5G A202006-100G A202006-25G
76691-214EA 10 USD
76691-214 77600-650 77600-652 77626-014 77626-016
Boc-Lys(Z)-OSu 96%
Boc-Lys(Z)-OSu

Precaución: for Research Use Only

Formula: C₂₃H₃₁N₃O₈
MDL Number: MFCD00037915
CAS Number: 34404-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR