cis-3-Hexenyl Butyrate 98%
Supplier: Ambeed
|
Warning
|
A201931-10G
A201931-5G
A201931-25G
76691-210EA
19.1
USD
76691-210
76691-212
77626-012
cis-3-Hexenyl Butyrate 98%
cis-3-Hexenyl Butyrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈O₂ MW: 170.25 g/mol |
MDL Number:
MFCD00036540 CAS Number: 16491-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...