4-Bromo-1-methyl-2-(methylsulfonyl)benzene 96%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-(methylsulphonyl)toluene

A201745-100MG A201745-1G A201745-250MG
76691-204EA 21.22 USD
76691-204 76691-206 76691-208
4-Bromo-1-methyl-2-(methylsulfonyl)benzene 96%
4-Bromo-1-methyl-2-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂S
MW: 249.13 g/mol
MDL Number: MFCD04037925
CAS Number: 254887-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR