2-Acetamido-3-(1H-imidazol-4-yl)propanoic acid hydrate 98%
Supplier: Ambeed
|
Warning
|
A200858-1G
A200858-25G
A200858-5G
76691-172EA
7.43
USD
76691-172
76691-174
76691-176
2-Acetamido-3-(1H-imidazol-4-yl)propanoic acid hydrate 98%
2-Acetamido-3-(1H-imidazol-4-yl)propanoic acid hydrate
Precaución: for Research Use Only
Formula:
C₈H₁₃N₃O₄ MW: 215.21 g/mol |
MDL Number:
MFCD00150969 CAS Number: 213178-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...