7-(Trifluoromethyl)-4-quinolinol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-7-(trifluoromethyl)quinoline

A200466-10G A200466-1G A200466-250MG A200466-5G
76691-160EA 150.65 USD
76691-160 76691-162 76691-164 76691-166
7-(Trifluoromethyl)-4-quinolinol 97%
7-(Trifluoromethyl)-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
MW: 213.1560 g/mol
MDL Number: MFCD00006779
CAS Number: 322-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR