3-Fluoro-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-o-tolualdehyde, 3-Fluoro-2-tolualdehyde, 3-Fluoro-o-tolylaldehyde

A200154-100G A200154-10G A200154-1G A200154-25G A200154-500G A200154-5G
76691-144EA 198 USD
76691-144 76691-146 76691-148 76691-150 76691-152 76691-154
3-Fluoro-2-methylbenzaldehyde 97%
3-Fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO
MDL Number: MFCD00075256
CAS Number: 147624-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR