8-(4-Chlorophenyl)-1,4-dioxaspiro[4.5]decane 95+%
Supplier: Ambeed
|
Warning
|
A200111-100MG
A200111-1G
A200111-250MG
76691-138EA
37
USD
76691-138
76691-140
76691-142
8-(4-Chlorophenyl)-1,4-dioxaspiro[4.5]decane 95+%
8-(4-Chlorophenyl)-1,4-dioxaspiro[4.5]decane
Precaución: for Research Use Only
Formula:
C₁₄H₁₇ClO₂ |
MDL Number:
MFCD07367861 CAS Number: 25253-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...