Bis(2-ethylhexyl) 3,4,5,6-tetrabromophthalate 95%

Supplier: Ambeed
Warning

A197473-100G A197473-10G A197473-25G A197473-500G A197473-5G
76691-082EA 139 USD
76691-082 76691-084 76691-086 76691-088 76691-090
Bis(2-ethylhexyl) 3,4,5,6-tetrabromophthalate 95%
Bis(2-ethylhexyl) 3,4,5,6-tetrabromophthalate

Precaución: for Research Use Only

Formula: C₂₄H₃₄Br₄O₄
MDL Number: MFCD01941091
CAS Number: 26040-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR