4,4'-Anisoin 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-1,2-bis(4-methoxyphenyl)ethanone, Anisoin

A196229-100G A196229-10G A196229-25G
76691-066EA 83 USD
76691-066 76691-068 76691-070
4,4'-Anisoin 97%
4,4'-Anisoin

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
MDL Number: MFCD00008411
CAS Number: 119-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR