8-Bromochroman-4-one 95%

Supplier: Ambeed
Warning

A195092-100MG A195092-1G A195092-250MG A195092-25G A195092-5G
76691-048EA 6 USD
76691-048 76691-050 76691-052 77567-832 77600-586
8-Bromochroman-4-one 95%
8-Bromochroman-4-one

Precaución: For research use only.

Formula: C₉H₇BrO₂
MDL Number: MFCD09744046
CAS Number: 204377-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR