N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Met-OSu, N-Boc-L-methionine N-Succinimidyl Ester
A194861-10G
A194861-25G
76691-036EA
25
USD
76691-036
76691-038
N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester 95%
N-(tert-Butoxycarbonyl)-L-methionine N-succinimidyl ester
Precaución: for Research Use Only
Formula:
C₁₄H₂₂N₂O₆S MW: 346.40 g/mol |
MDL Number:
MFCD00042760 CAS Number: 3845-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...