4'-Bromo-2'-fluoroacetanilide 98%

Supplier: Ambeed
Warning

A194505-100G A194505-25G A194505-5G
76691-016EA 59 USD
76691-016 76691-018 76691-020
4'-Bromo-2'-fluoroacetanilide 98%
4'-Bromo-2'-fluoroacetanilide

Precaución: for Research Use Only

Formula: H₃CC(O)NH(C₆H₃BrF)
MDL Number: MFCD00042588
CAS Number: 326-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR