Anthracen-9-ylmethanamine 97%

Supplier: Ambeed
Warning

A194260-100MG A194260-1G A194260-250MG
76690-994EA 20 USD
76690-994 76690-996 76690-998
Anthracen-9-ylmethanamine 97%
Anthracen-9-ylmethanamine

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
MDL Number: MFCD06212577
CAS Number: 2476-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR